(1S,2R,4AS,6aS,6bR,8aR,10S,12aR,12bR,14bS)-10-acetoxy-1,2,6a,6b,9,9,12a-heptamethyl-1,3,4,5,6,6a,6b,7,8,8a,9,10,11,12,12a,12b,13,14b-octadecahydropicene-4a(2H)-carboxylic acid 95%

Supplier: Ambeed
Warning

A260121-2MG A260121-5MG
76827-116EA 119.89 USD
76827-116 76827-118
(1S,2R,4AS,6aS,6bR,8aR,10S,12aR,12bR,14bS)-10-acetoxy-1,2,6a,6b,9,9,12a-heptamethyl-1,3,4,5,6,6a,6b,7,8,8a,9,10,11,12,12a,12b,13,14b-octadecahydropicene-4a(2H)-carboxylic acid 95%
(1S,2R,4AS,6aS,6bR,8aR,10S,12aR,12bR,14bS)-10-acetoxy-1,2,6a,6b,9,9,12a-heptamethyl-1,3,4,5,6,6a,6b,7,8,8a,9,10,11,12,12a,12b,13,14b-octadecahydropicene-4a(2H)-carboxylic acid

Precaución: for Research Use Only

Formula: C₃₂H₅₀O₄
MW: 498.74 g/mol
MDL Number: MFCD09752431
CAS Number: 7372-30-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR