3-(Ethoxycarbonylmethyl)phenylboronic acid 95%

Supplier: Ambeed
Warning

Synonyms: (3-(2-Ethoxy-2-oxoethyl)phenyl)boronic acid

A257556-1G A257556-250MG
76827-088EA 28.65 USD
76827-088 77885-675 77885-676
3-(Ethoxycarbonylmethyl)phenylboronic acid 95%
3-(Ethoxycarbonylmethyl)phenylboronic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₃BO₄
MDL Number: MFCD08704681
CAS Number: 1256345-69-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR