Dibenzo[f,h]quinoxaline 99+%

Supplier: Ambeed
Warning

A256887-1G A256887-250MG A256887-5G
76827-076EA 76.39 USD
76827-076 76827-078 76827-080
Dibenzo[f,h]quinoxaline 99+%
Dibenzo[f,h]quinoxaline

Precaución: for Research Use Only

Formula: C₁₆H₁₀N₂
MW: 230.26 g/mol
MDL Number: MFCD00114919
CAS Number: 217-68-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR