4,4'-Biphenyldiboronic acid bis(pinacol)ester 97%

Supplier: Ambeed
Warning

Synonyms: 4,4,5,5-Tetramethyl-2-{4-[4-(tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]phenyl}-1,3,2-dioxaborolane, 4,4'-Bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1,1'-biphenyl

A255470-10G A255470-1G A255470-250MG A255470-5G
76827-048EA 87 USD
76827-048 76827-050 76827-052 77683-775
4,4'-Biphenyldiboronic acid bis(pinacol)ester 97%
4,4'-Biphenyldiboronic acid bis(pinacol) ester

Precaución: For research use only.

Formula: C₂₄H₃₂B₂O₄
MW: 406.14 g/mol
MDL Number: MFCD08669545
CAS Number: 207611-87-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR