1-tert-Butylimidazole 98% (by GC)

Supplier: Ambeed
Warning

A254641-1G A254641-250MG A254641-5G A254641-25G
76827-042EA 30.77 USD
76827-042 76827-044 76827-046 77550-618
1-tert-Butylimidazole 98% (by GC)
1-tert-Butylimidazole

Precaución: for Research Use Only

Formula: C₇H₁₂N₂
MW: 124.19 g/mol
MDL Number: MFCD19441219
CAS Number: 45676-04-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR