(6-Hydroxyspiro[3.3]hept-2-yl)carbamicacid-tert-butyl ester 97%

Supplier: Ambeed
Warning

Synonyms: tert-butyl N-{6-hydroxyspiro[3.3]heptan-2-yl}carbamate, 2-(Boc-amino)-6-hydroxyspiro[3.3]heptane, tert-Butyl (6-hydroxyspiro[3.3]heptan-2-yl)carbamate

A254594-100MG A254594-250MG A254594-1G A254594-5G
76827-032EA 31.83 USD
76827-032 76827-034 77612-444 77683-767
(6-Hydroxyspiro[3.3]hept-2-yl)carbamicacid-tert-butyl ester 97%
(6-Hydroxyspiro[3.3]hept-2-yl)carbamicacid-tert-butyl ester

Precaución: For research use only.

Formula: C₁₂H₂₁NO₃
MW: 227.31 g/mol
MDL Number: MFCD09702342
CAS Number: 1000933-99-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR