6-Bromocinnolin-4-ol 98%

Supplier: Ambeed
Warning

A254470-250MG A254470-5G A254470-1G
76827-028EA 40.32 USD
76827-028 76827-030 77683-763
6-Bromocinnolin-4-ol 98%
6-Bromocinnolin-4-ol

Precaución: For research use only.

Formula: C₈H₅BrN₂O
MW: 225.04 g/mol
MDL Number: MFCD09834642
CAS Number: 876-88-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR