SB-242084 Dihydrochloride 97%

Supplier: Ambeed
Warning

A253004-1MG A253004-5MG
76827-020EA 14 USD
76827-020 76827-022
SB-242084 Dihydrochloride 97%
SB-242084 Dihydrochloride

Precaución: for Research Use Only

Formula: C₂₁H₂₁Cl₃N₄O₂
MDL Number: MFCD02684417
CAS Number: 1049747-87-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR