5-Bromothieno[2,3-d]pyrimidin-4(3H)-one 95%

Supplier: Ambeed
Warning

A252668-100MG A252668-1G A252668-250MG A252668-5G
76827-002EA 27.59 USD
76827-002 76827-004 76827-006 76827-008
5-Bromothieno[2,3-d]pyrimidin-4(3H)-one 95%
5-Bromothieno[2,3-d]pyrimidin-4(3H)-one

Precaución: for Research Use Only

Formula: C₆H₃BrN₂OS
MW: 231.07 g/mol
MDL Number: MFCD25542166
CAS Number: 1239460-83-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR