4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)dibenzothiophene 98%

Supplier: Ambeed
Warning

Synonyms: 2-(Dibenzo[b,d]thiophen-4-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-(Dibenzothiophene-4-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 4-Dibenzothiopheneboronic acid Pinacol ester

A252107-1G A252107-250MG A252107-25G A252107-5G
76826-980EA 21.22 USD
76826-980 76826-982 76826-984 76826-986
4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)dibenzothiophene 98%
4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)dibenzothiophene

Precaución: for Research Use Only

Formula: C₁₈H₁₉BO₂S
MW: 310.22 g/mol
MDL Number: MFCD18452170
CAS Number: 912824-84-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR