2-(4-Hydroxyphenyl)benzo[b]thiophene-6-ol 98%

Supplier: Ambeed
Warning

Synonyms: 2-(4-Hydroxyphenyl)benzo[b]thiophen-6-ol

A251043-10MG A251043-1MG A251043-25MG A251043-50MG A251043-5MG
76826-954EA 74.27 USD
76826-954 76826-956 76826-958 76826-960 76826-962
2-(4-Hydroxyphenyl)benzo[b]thiophene-6-ol 98%
2-(4-Hydroxyphenyl)benzo[b]thiophene-6-ol

Precaución: for Research Use Only

Formula: C₁₄H₁₀O₂S
MW: 242.29 g/mol
MDL Number: MFCD16619490
CAS Number: 63676-22-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR