1-(4-Bromophenyl)-2-methyl-1-propanone 98%

Supplier: Ambeed
Warning

Synonyms: 1-(4-Bromophenyl)-2-methylpropan-1-one

A251016-100MG A251016-1G A251016-250MG A251016-5G
76826-946EA 24.41 USD
76826-946 76826-948 76826-950 76826-952
1-(4-Bromophenyl)-2-methyl-1-propanone 98%
1-(4-Bromophenyl)-2-methyl-1-propanone

Precaución: for Research Use Only

Formula: C₁₀H₁₁BrO
MW: 227.10 g/mol
MDL Number: MFCD09891214
CAS Number: 49660-93-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR