3-(4-Morpholinylcarbonyl)phenylboronic acid pinacol ester 98%

Supplier: Ambeed
Warning

Synonyms: Morpholino(3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)methanone, 3-(4-Morpholinylcarbonyl)benzeneboronic acid pinacol ester

A250298-1G A250298-25G A250298-5G
76826-932EA 30.77 USD
76826-932 76826-934 76826-936
3-(4-Morpholinylcarbonyl)phenylboronic acid pinacol ester 98%
3-(4-Morpholinylcarbonyl)phenylboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₇H₂₄BNO₄
MW: 317.19 g/mol
MDL Number: MFCD09266172
CAS Number: 1036991-25-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR