5-Chlorothieno[3,2-b]pyridine 95%

Supplier: Ambeed
Warning

A250148-100MG A250148-1G A250148-250MG
76826-926EA 41.38 USD
76826-926 76826-928 76826-930
5-Chlorothieno[3,2-b]pyridine 95%
5-Chlorothieno[3,2-b]pyridine

Precaución: for Research Use Only

Formula: C₇H₄ClNS
MW: 169.631 g/mol
MDL Number: MFCD09909817
CAS Number: 65977-55-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR