1,10-Phenanthroline-4,7-dicarboxylic acid 95%

Supplier: Ambeed
Warning

A250014-100MG A250014-250MG
76826-920EA 150 USD
76826-920 76826-922
1,10-Phenanthroline-4,7-dicarboxylic acid 95%
1,10-Phenanthroline-4,7-dicarboxylic acid

Precaución: for Research Use Only

Formula: C₁₄H₈N₂O₄
MDL Number: MFCD19441391
CAS Number: 31301-31-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR