Ethyl 3-(2-fluoro-4-methoxyphenyl)-3-oxopropanoate 95+%

Supplier: Ambeed
Warning

A249221-1G A249221-250MG
76826-916EA 476.35 USD
76826-916 76826-918
Ethyl 3-(2-fluoro-4-methoxyphenyl)-3-oxopropanoate 95+%
Ethyl 3-(2-fluoro-4-methoxyphenyl)-3-oxopropanoate

Precaución: for Research Use Only

Formula: C₁₂H₁₃FO₄
MW: 240.22 g/mol
MDL Number: MFCD08702766
CAS Number: 188182-79-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR