Bis(4-methoxyphenyl)phosphine oxide 95%

Supplier: Ambeed
Warning

A248959-1G A248959-250MG A248959-5G A248959-25G
76826-910EA 30 USD
76826-910 76826-912 76826-914 77612-400
Bis(4-methoxyphenyl)phosphine oxide 95%
Bis(4-methoxyphenyl)phosphine oxide

Precaución: for Research Use Only

Formula: C₁₄H₁₅O₃P
MDL Number: MFCD06754095
CAS Number: 15754-51-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR