(5-(Pyridin-3-yl)-1H-pyrazol-3-yl)methanol 97%

Supplier: Ambeed
Warning

A248480-100MG A248480-1G A248480-250MG
76826-898EA 41.38 USD
76826-898 76826-900 76826-902
(5-(Pyridin-3-yl)-1H-pyrazol-3-yl)methanol 97%
(5-(Pyridin-3-yl)-1H-pyrazol-3-yl)methanol

Precaución: for Research Use Only

Formula: C₉H₉N₃O
MW: 175.19 g/mol
MDL Number: MFCD20527516
CAS Number: 287494-03-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR