2-Ethoxy-1-fluoro-4-nitrobenzene 97%

Supplier: Ambeed
Warning

A248441-1G A248441-250MG
76826-894EA 186.72 USD
76826-894 76826-896
2-Ethoxy-1-fluoro-4-nitrobenzene 97%
2-Ethoxy-1-fluoro-4-nitrobenzene

Precaución: for Research Use Only

Formula: C₈H₈FNO₃
MW: 185.152 g/mol
MDL Number: MFCD20133797
CAS Number: 1093656-34-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR