2-Acetamido-6-aminopyridine 98%

Supplier: Ambeed
Warning

Synonyms: N-(6-Aminopyridin-2-yl)acetamide

A247227-100G A247227-1G A247227-25G A247227-5G
76826-860EA 314.03 USD
76826-860 76826-862 76826-864 76826-866
2-Acetamido-6-aminopyridine 98%
2-Acetamido-6-aminopyridine

Precaución: for Research Use Only

Formula: C₇H₉N₃O
MW: 151.17 g/mol
MDL Number: MFCD01099052
CAS Number: 1075-62-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR