4-Iodo-7H-pyrrolo[2,3-d]pyrimidine 95%

Supplier: Ambeed
Warning

A247102-10G A247102-1G A247102-250MG A247102-5G
76826-852EA 594.11 USD
76826-852 76826-854 76826-856 76826-858
4-Iodo-7H-pyrrolo[2,3-d]pyrimidine 95%
4-Iodo-7H-pyrrolo[2,3-d]pyrimidine

Precaución: for Research Use Only

Formula: C₆H₄IN₃
MW: 245.02 g/mol
MDL Number: MFCD17016136
CAS Number: 1100318-96-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR