Di-tert-butyl(2',4',6'-triisopropyl-3-methoxy-6-methyl-[1,1'-biphenyl]-2-yl)phosphine 98%
Supplier: Ambeed
|
Warning
|
A246598-100MG
A246598-1G
A246598-250MG
A246598-50MG
76826-842EA
58
USD
76826-842
76826-844
76826-846
76826-848
Di-tert-butyl(2',4',6'-triisopropyl-3-methoxy-6-methyl-[1,1'-biphenyl]-2-yl)phosphine 98%
Di-tert-butyl(2',4',6'-triisopropyl-3-methoxy-6-methyl-[1,1'-biphenyl]-2-yl)phosphine
Precaución: for Research Use Only
Formula:
C₃₁H₄₉OP MW: 468.69 g/mol |
MDL Number:
MFCD20922895 CAS Number: 1262046-34-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...