3,5-Dimethylcyclohexanol (mixture of isomers) 98%

Supplier: Ambeed
Warning

A245553-100G A245553-1G A245553-25G A245553-5G
76826-824EA 431 USD
76826-824 76826-826 76826-828 76826-830
3,5-Dimethylcyclohexanol (mixture of isomers) 98%
3,5-Dimethylcyclohexanol (mixture of isomers)

Precaución: for Research Use Only

Formula: C₈H₁₆O
MDL Number: MFCD00001447
CAS Number: 5441-52-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR