2-Hydroxyisobutyramide 97%

Supplier: Ambeed
Warning

Synonyms: 2-Hydroxy-2-methylpropanamide, 2-Hydroxy-2-methylpropionamide

A244732-100G A244732-10G A244732-1G A244732-5G
76826-808EA 314.03 USD
76826-808 76826-810 76826-812 76826-814
2-Hydroxyisobutyramide 97%
2-Hydroxyisobutyramide

Precaución: for Research Use Only

Formula: C₄H₉NO₂
MW: 103.12 g/mol
MDL Number: MFCD08063365
CAS Number: 13027-88-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR