2,4-Dihydroxy-5-nitrobenzoic acid 95%

Supplier: Ambeed
Warning

A243554-10G A243554-1G A243554-250MG A243554-5G
76826-788EA 159.14 USD
76826-788 76826-790 76826-792 76826-794
2,4-Dihydroxy-5-nitrobenzoic acid 95%
2,4-Dihydroxy-5-nitrobenzoic acid

Precaución: for Research Use Only

Formula: C₇H₅NO₆
MW: 199.11 g/mol
MDL Number: MFCD08693128
CAS Number: 13722-96-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR