Triphenylmethylium tetrakis(pentafluorophenyl)borate 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
Trityltetra(pentafluorophenyl)borate, Triphenylmethylium tetrakis(perfluorophenyl)borate, Trityl tetrakis(pentafluorophenyl)borate, Triphenylcarbenium Tetrakis(pentafluorophenyl)borate, Tritylium Tetrakis(pentafluorophenyl)borate
A243475-1G
A243475-250MG
A243475-25G
A243475-5G
A243475-100G
76826-776EA
10
USD
76826-776
76826-778
76826-780
76826-782
77612-352
Triphenylmethylium tetrakis(pentafluorophenyl)borate 97%
Triphenylmethylium tetrakis(pentafluorophenyl)borate
Precaución: for Research Use Only
Formula:
C₄₃H₁₅BF₂₀ MW: 922.37 g/mol |
MDL Number:
MFCD03426981 CAS Number: 136040-19-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...