3-Fluoro-5-nitrobenzyl chloride 95%

Supplier: Ambeed
Danger

Synonyms: 1-(Chloromethyl)-3-fluoro-5-nitrobenzene

A243191-10G A243191-1G A243191-25G A243191-5G
76826-764EA 274.78 USD
76826-764 76826-766 76826-768 76826-770
3-Fluoro-5-nitrobenzyl chloride 95%
3-Fluoro-5-nitrobenzyl chloride

Precaución: for Research Use Only

Formula: C₇H₅ClFNO₂
MW: 189.572 g/mol
MDL Number: MFCD13185626
CAS Number: 1214344-25-8
UN: 3261
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR