5-Bromo-2-chloro-4-(trifluoromethyl)pyrimidine 98%

Supplier: Ambeed
Warning

Synonyms: 5-Bromo-2-chloro-4-(trifluoromethyl)-1,3-diazine

A242878-100MG A242878-1G A242878-250MG
76826-758EA 35.01 USD
76826-758 76826-760 76826-762
5-Bromo-2-chloro-4-(trifluoromethyl)pyrimidine 98%
5-Bromo-2-chloro-4-(trifluoromethyl)pyrimidine

Precaución: for Research Use Only

Formula: C₅HBrClF₃N₂
MW: 261.427 g/mol
MDL Number: MFCD13176600
CAS Number: 785777-92-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR