(S)-tert-Butyl 3-(2-ethoxy-2-oxoethyl)morpholine-4-carboxylate 98%

Supplier: Ambeed
Warning

A242030-100MG A242030-1G A242030-250MG
76826-750EA 105.03 USD
76826-750 76826-752 76826-754
(S)-tert-Butyl 3-(2-ethoxy-2-oxoethyl)morpholine-4-carboxylate 98%
(S)-tert-Butyl 3-(2-ethoxy-2-oxoethyl)morpholine-4-carboxylate

Precaución: for Research Use Only

Formula: C₁₃H₂₃NO₅
MW: 273.32 g/mol
MDL Number: MFCD23701548
CAS Number: 761460-04-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR