(4,5-Dimethylthiazol-2-yl)methanol 95%

Supplier: Ambeed
Warning

A242012-100MG A242012-1G A242012-250MG A242012-5G
76826-742EA 26.53 USD
76826-742 76826-744 76826-746 76826-748
(4,5-Dimethylthiazol-2-yl)methanol 95%
(4,5-Dimethylthiazol-2-yl)methanol

Precaución: for Research Use Only

Formula: C₆H₉NOS
MW: 143.207 g/mol
MDL Number: MFCD12198452
CAS Number: 99839-16-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR