Methyl 3-(((tert-butoxycarbonyl)amino)methyl)benzoate 97%

Supplier: Ambeed
Warning

A241818-100MG A241818-1G A241818-250MG A241818-5G
76826-734EA 8.49 USD
76826-734 76826-736 76826-738 76826-740
Methyl 3-(((tert-butoxycarbonyl)amino)methyl)benzoate 97%
Methyl 3-(((tert-butoxycarbonyl)amino)methyl)benzoate

Precaución: for Research Use Only

Formula: C₁₄H₁₉NO₄
MW: 265.31 g/mol
MDL Number: MFCD14583094
CAS Number: 180863-55-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR