5-Nitro-2-(trifluoromethyl)benzoic acid 97%

Supplier: Ambeed
Warning

A241500-1G A241500-250MG A241500-5G
76826-728EA 65.78 USD
76826-728 76826-730 76826-732
5-Nitro-2-(trifluoromethyl)benzoic acid 97%
5-Nitro-2-(trifluoromethyl)benzoic acid

Precaución: for Research Use Only

Formula: C₈H₄F₃NO₄
MW: 235.12 g/mol
MDL Number: MFCD13185777
CAS Number: 847547-06-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR