2,4-Dibromo-5-methylthiazole 98%

Supplier: Ambeed
Warning

A241488-100MG A241488-250MG A241488-5G
76826-724EA 10 USD
76826-724 76826-726 77550-558
2,4-Dibromo-5-methylthiazole 98%
2,4-Dibromo-5-methylthiazole

Precaución: for Research Use Only

Formula: C₄H₃Br₂NS
MDL Number: MFCD23135672
CAS Number: 1206708-88-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR