1-(3-Hydroxy-2-nitrophenyl)ethanone 98%

Supplier: Ambeed
Warning

A240418-100MG A240418-10G A240418-1G A240418-250MG A240418-25G A240418-5G
76826-706EA 5.31 USD
76826-706 76826-708 76826-710 76826-712 76826-714 76826-716
1-(3-Hydroxy-2-nitrophenyl)ethanone 98%
1-(3-Hydroxy-2-nitrophenyl)ethanone

Precaución: for Research Use Only

Formula: C₈H₇NO₄
MW: 181.15 g/mol
MDL Number: MFCD06251784
CAS Number: 53967-72-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR