tert-Butyl 3-(phenylamino)pyrrolidine-1-carboxylate 95%

Supplier: Ambeed
Warning

A240080-100MG A240080-1G A240080-250MG
76826-700EA 27.59 USD
76826-700 76826-702 76826-704
tert-Butyl 3-(phenylamino)pyrrolidine-1-carboxylate 95%
tert-Butyl 3-(phenylamino)pyrrolidine-1-carboxylate

Precaución: for Research Use Only

Formula: C₁₅H₂₂N₂O₂
MW: 262.34 g/mol
MDL Number: MFCD09608043
CAS Number: 816468-24-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR