2-Fluoro-6-methyl-3-pyridineboronic acid pinacol ester 98%

Supplier: Ambeed
Warning

Synonyms: 2-Fluoro-6-methyl-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine

A239990-100MG A239990-250MG
76826-692EA 31.83 USD
76826-692 76826-694
2-Fluoro-6-methyl-3-pyridineboronic acid pinacol ester 98%
2-Fluoro-6-methyl-3-pyridineboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₂H₁₇BFNO₂
MW: 237.078 g/mol
MDL Number: MFCD08063091
CAS Number: 1218790-38-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR