Ethyl 2-chloro-5-methylnicotinate 98%

Supplier: Ambeed
Warning

A238502-100MG A238502-1G
76826-668EA 39.26 USD
76826-668 76826-670
Ethyl 2-chloro-5-methylnicotinate 98%
Ethyl 2-chloro-5-methylnicotinate

Precaución: for Research Use Only

Formula: C₉H₁₀ClNO₂
MW: 199.63 g/mol
MDL Number: MFCD16610952
CAS Number: 894074-85-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR