Methyl-2-fluoro-5-methylbenzoate 98%

Supplier: Ambeed
Warning

A238455-100G A238455-10G
76826-666EA 187 USD
76826-666 77885-610
Methyl-2-fluoro-5-methylbenzoate 98%
Methyl 2-fluoro-5-methylbenzoate

Precaución: for Research Use Only

Formula: C₉H₉FO₂
MDL Number: MFCD06203924
CAS Number: 2967-93-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR