Ethyl 4-oxotetrahydrofuran-3-carboxylate 97%

Supplier: Ambeed
Warning

A237327-250MG A237327-1G
76826-650EA 23 USD
76826-650 77612-310
Ethyl 4-oxotetrahydrofuran-3-carboxylate 97%
Ethyl 4-oxotetrahydrofuran-3-carboxylate

Precaución: for Research Use Only

Formula: C₇H₁₀O₄
MDL Number: MFCD16038834
CAS Number: 89898-51-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR