(4-(((Benzyloxy)carbonyl)(ethyl)amino)phenyl)boronic acid 98%

Supplier: Ambeed
Warning

A237037-100MG A237037-1G A237037-250MG
76826-632EA 11 USD
76826-632 76826-634 76826-636
(4-(((Benzyloxy)carbonyl)(ethyl)amino)phenyl)boronic acid 98%
(4-(((Benzyloxy)carbonyl)(ethyl)amino)phenyl)boronic acid

Precaución: for Research Use Only

Formula: C₁₆H₁₈BNO₄
MDL Number: MFCD21609545
CAS Number: 1221448-69-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR