5-Amino-2-(piperidin-1-yl)benzonitrile 95%

Supplier: Ambeed
Warning

A236948-100MG A236948-10G A236948-1G A236948-250MG
76826-620EA 41.38 USD
76826-620 76826-622 76826-624 76826-626
5-Amino-2-(piperidin-1-yl)benzonitrile 95%
5-Amino-2-(piperidin-1-yl)benzonitrile

Precaución: for Research Use Only

Formula: C₁₂H₁₅N₃
MW: 201.26 g/mol
MDL Number: MFCD14617260
CAS Number: 34595-33-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR