3-(5,5-Dimethyl-1,3,2-dioxaborinan-2-yl)aniline hydrochloride 95+%

Supplier: Ambeed
Warning

A236757-1G A236757-250MG A236757-5G
76826-608EA 22 USD
76826-608 76826-610 76826-612
3-(5,5-Dimethyl-1,3,2-dioxaborinan-2-yl)aniline hydrochloride 95+%
3-(5,5-Dimethyl-1,3,2-dioxaborinan-2-yl)aniline hydrochloride

Precaución: for Research Use Only

Formula: C₁₁H₁₇BClNO₂
MW: 241.52 g/mol
MDL Number: MFCD06659912
CAS Number: 850567-43-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR