1-(3-Chlorophenyl)cyclopropanecarboxylic acid 95%

Supplier: Ambeed
Warning

A236494-1G A236494-250MG A236494-5G
76826-598EA 23 USD
76826-598 76826-600 76826-602
1-(3-Chlorophenyl)cyclopropanecarboxylic acid 95%
1-(3-Chlorophenyl)cyclopropanecarboxylic acid

Precaución: for Research Use Only

Formula: C₁₀H₉ClO₂
MDL Number: MFCD07374442
CAS Number: 124276-34-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR