6-Bromo-1-hydroxynaphthalene 98%

Supplier: Ambeed
Warning

A235854-100MG A235854-1G A235854-250MG
76826-586EA 80.63 USD
76826-586 76826-588 76826-590
6-Bromo-1-hydroxynaphthalene 98%
6-Bromo-1-hydroxynaphthalene

Precaución: for Research Use Only

Formula: C₁₀H₇BrO
MW: 223.06 g/mol
MDL Number: MFCD15528811
CAS Number: 91270-68-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR