1,2-Dibromo-4,5-difluoro-3-nitrobenzene 97%

Supplier: Ambeed
Warning

A235361-1G A235361-5G
76826-582EA 32 USD
76826-582 76826-584
1,2-Dibromo-4,5-difluoro-3-nitrobenzene 97%
1,2-Dibromo-4,5-difluoro-3-nitrobenzene

Precaución: for Research Use Only

Formula: C₆HBr₂F₂NO₂
MDL Number: MFCD11226312
CAS Number: 1481-57-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR