4-(4-((4'-Chloro-4,4-dimethyl-3,4,5,6-tetrahydro-[1,1'-biphenyl]-2-yl)methyl)piperazin-1-yl)benzoic acid 95%

Supplier: Ambeed
Warning

A235146-100MG A235146-250MG A235146-1G
76826-572EA 63.66 USD
76826-572 76826-574 77593-512
4-(4-((4'-Chloro-4,4-dimethyl-3,4,5,6-tetrahydro-[1,1'-biphenyl]-2-yl)methyl)piperazin-1-yl)benzoic acid 95%
4-(4-((4'-Chloro-4,4-dimethyl-3,4,5,6-tetrahydro-[1,1'-biphenyl]-2-yl)methyl)piperazin-1-yl)benzoic acid

Precaución: for Research Use Only

Formula: C₂₆H₃₁ClN₂O₂
MW: 438.99 g/mol
MDL Number: MFCD16251302
CAS Number: 1044598-91-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR