1-Phenyl-3-(trifluoromethyl)pyrazole 95%

Supplier: Ambeed
Warning

Synonyms: 1-Phenyl-3-(trifluoromethyl)-1H-pyrazole

A234793-100MG A234793-1G A234793-250MG
76826-566EA 41.38 USD
76826-566 76826-568 76826-570
1-Phenyl-3-(trifluoromethyl)pyrazole 95%
1-Phenyl-3-(trifluoromethyl)pyrazole

Precaución: for Research Use Only

Formula: C₁₀H₇F₃N₂
MW: 212.171 g/mol
MDL Number: MFCD03094197
CAS Number: 99498-65-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR