(S)-2-(2,6-Dichlorobenzamido)-3-(2',6'-dimethoxy-[1,1'-biphenyl]-4-yl)propanoic acid 98%

Supplier: Ambeed
Warning

Synonyms: TR-14035

A234495-10MG A234495-1MG A234495-2MG A234495-50MG A234495-5MG
76826-554EA 48.81 USD
76826-554 76826-556 76826-558 76826-560 76826-562
(S)-2-(2,6-Dichlorobenzamido)-3-(2',6'-dimethoxy-[1,1'-biphenyl]-4-yl)propanoic acid 98%
(S)-2-(2,6-Dichlorobenzamido)-3-(2',6'-dimethoxy-[1,1'-biphenyl]-4-yl)propanoic acid

Precaución: for Research Use Only

Formula: C₂₄H₂₁Cl₂NO₅
MW: 474.33 g/mol
MDL Number: MFCD09970400
CAS Number: 232271-19-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR