Diethyl (2-amino-2-oxoethyl)phosphonate 95%

Supplier: Ambeed
Warning

A234328-1G A234328-25G A234328-5G
76826-548EA 29.71 USD
76826-548 76826-550 76826-552
Diethyl (2-amino-2-oxoethyl)phosphonate 95%
Diethyl (2-amino-2-oxoethyl)phosphonate

Precaución: for Research Use Only

Formula: C₆H₁₄NO₄P
MW: 195.15 g/mol
MDL Number: MFCD00014805
CAS Number: 5464-68-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR