(4-Methoxy-3-nitrophenyl)methanol 95%

Supplier: Ambeed
Warning

A233689-1G A233689-250MG A233689-5G
76826-538EA 33 USD
76826-538 76826-540 76826-542
(4-Methoxy-3-nitrophenyl)methanol 95%
(4-Methoxy-3-nitrophenyl)methanol

Precaución: for Research Use Only

Formula: C₈H₉NO₄
MDL Number: MFCD02683526
CAS Number: 41870-24-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR